Assay Design



Gene Transcripts

Gene Symbol
Rbp1
Entrez Gene ID
19659
Gene Name
retinol binding protein 1, cellular
Gene Aliases
CRBPI, Crbp, Rbp-1
Chromosome Location
Chr.9: 98422961 - 98446551 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011254.5 NP_035384.1 3 - 4 2369 72
GenBank mRNA AK011641.1 - 3 - 4 517 72
BC018254.1 - 3 - 4 496 72
X60367.1 - 3 - 4 2368 72

View Full Product Details