Assay Design



Gene Transcripts

Gene Symbol
Tbx6
Entrez Gene ID
21389
Gene Name
T-box 6
Gene Aliases
rv
Chromosome Location
Chr.7: 126781483 - 126785548 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011538.2 NP_035668.2 6 - 7 942 69
GenBank mRNA AY654733.1 - 6 - 7 918 69
U57331.1 - 6 - 7 942 69

View Full Product Details