Assay Design



Gene Transcripts

Gene Symbol
Tcf12
Entrez Gene ID
21406
Gene Name
transcription factor 12
Gene Aliases
A130037E08Rik, ALF1, HEB, HEBAlt, HTF-4, HTF4, ME1, REB, bHLHb20
Chromosome Location
Chr.9: 71844252 - 72111819 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001253862.1 NP_001240791.1 9 - 10 893 72
NM_001253863.1 NP_001240792.1 2 - 3 272 72
NM_001253864.1 NP_001240793.1 2 - 3 272 72
NM_001253865.1 NP_001240794.1 4 - 5 519 72
NM_011544.3 NP_035674.2 9 - 10 950 72
GenBank mRNA AK078415.1 - 2 - 3 265 72
AK132002.1 - 2 - 3 257 72
AK133958.1 - 2 - 3 260 72
AK134037.1 - 9 - 10 894 72
AK135376.1 - 2 - 3 240 72
BC037097.1 - 4 - 5 507 72
CJ164494.1 - 2 - 3 274 72
M97635.1 - 9 - 10 921 72
X64840.1 - 9 - 10 750 72

View Full Product Details