Assay Design



Gene Transcripts

Gene Symbol
Cd40lg
Entrez Gene ID
21947
Gene Name
CD40 ligand
Gene Aliases
CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39
Chromosome Location
Chr.X: 57212143 - 57224042 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011616.2 NP_035746.2 1 - 2 162 118
GenBank mRNA BC119225.1 - 1 - 2 209 118
EU274582.1 - 1 - 2 150 118
X65453.2 - 1 - 2 162 118

View Full Product Details