Assay Design



Gene Transcripts

Gene Symbol
Twist1
Entrez Gene ID
22160
Gene Name
twist basic helix-loop-helix transcription factor 1
Gene Aliases
AA960487, M-Twist, Pde, Ska10, Ska<m10Jus>, Twist, bHLHa38, pdt
Chromosome Location
Chr.12: 33957671 - 33959831 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011658.2 NP_035788.1 1 - 2 965 113
GenBank mRNA BC033434.1 - 1 - 2 791 113
BC083139.1 - 1 - 2 965 113
M63650.1 - 1 - 2 824 113

View Full Product Details