Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011850.2 | NP_035980.1 | 1 - 2 | 579 | 73 |
GenBank mRNA | AK004930.1 | - | 1 - 2 | 589 | 73 |
AY360323.1 | - | 1 - 2 | 544 | 73 | |
AY360324.1 | - | 1 - 2 | 544 | 73 | |
BC019540.1 | - | 1 - 2 | 579 | 73 |