Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011910.2 | NP_036040.1 | 3 - 4 | 294 | 91 |
GenBank mRNA | AF172175.1 | - | 3 - 4 | 294 | 91 |
AY065984.1 | - | 3 - 4 | 258 | 91 | |
BC107299.1 | - | 3 - 4 | 282 | 91 | |
BC107300.1 | - | 3 - 4 | 280 | 91 |