Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011915.2 | NP_036045.1 | 6 - 7 | 1064 | 63 |
GenBank mRNA | AF122923.1 | - | 6 - 7 | 838 | 63 |
AK077698.1 | - | 6 - 7 | 1064 | 63 | |
AK142443.1 | - | 6 - 7 | 1065 | 63 | |
AK158359.1 | - | 6 - 7 | 1065 | 63 | |
AK207348.1 | - | 4 - 5 | 348 | 63 | |
BC004048.1 | - | 2 - 3 | 344 | 63 | |
BC013268.1 | - | 6 - 7 | 865 | 63 |