Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013521.2 | NP_038549.1 | 2 - 2 | 217 | 100 |
GenBank mRNA | AK137714.1 | - | 2 - 2 | 219 | 100 |
BY194974.1 | - | 2 - 2 | 220 | 100 | |
BY750286.1 | - | 2 - 2 | 220 | 100 |