Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039195.1 | NP_001034284.1 | 3 - 4 | 875 | 67 |
NM_001083806.1 | NP_001077275.1 | 3 - 4 | 875 | 67 | |
NM_013540.2 | NP_038568.2 | 3 - 4 | 875 | 67 | |
GenBank mRNA | AB109210.1 | - | 3 - 4 | 468 | 67 |
AB109211.1 | - | 3 - 4 | 468 | 67 | |
AB111957.1 | - | 3 - 4 | 468 | 67 | |
AK014389.1 | - | 3 - 4 | 920 | 67 | |
AK043490.1 | - | 3 - 4 | 848 | 67 | |
AK044574.1 | - | 3 - 4 | 807 | 67 | |
AK046861.1 | - | 3 - 4 | 875 | 67 | |
AK086614.1 | - | 3 - 4 | 809 | 67 | |
AK134572.1 | - | 3 - 4 | 806 | 67 | |
AK134647.1 | - | 2 - 3 | 321 | 67 | |
AK134772.1 | - | 2 - 3 | 446 | 67 | |
AK141525.1 | - | 3 - 4 | 850 | 67 | |
AK163941.1 | - | 3 - 4 | 868 | 67 | |
BC048248.1 | - | 3 - 4 | 734 | 67 | |
BC066193.1 | - | 3 - 4 | 663 | 67 | |
X57498.1 | - | 3 - 4 | 670 | 67 |