Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001244198.1 | NP_001231127.1 | 12 - 13 | 1514 | 97 |
NM_001244200.1 | NP_001231129.1 | 13 - 14 | 1726 | 97 | |
NM_001244201.1 | NP_001231130.1 | 11 - 12 | 1472 | 97 | |
NM_001244202.1 | NP_001231131.1 | 11 - 12 | 2028 | 97 | |
NM_013627.5 | NP_038655.1 | 12 - 13 | 2070 | 97 | |
GenBank mRNA | AF457141.1 | - | 6 - 7 | 793 | 97 |
AF457142.1 | - | 7 - 8 | 610 | 97 | |
AJ292077.1 | - | 11 - 12 | 1331 | 97 | |
AJ307468.1 | - | 11 - 12 | 1338 | 97 | |
AK028059.1 | - | 11 - 12 | 1473 | 97 | |
AK032616.1 | - | 12 - 13 | 1514 | 97 | |
AK045805.1 | - | 11 - 12 | 2030 | 97 | |
AK139054.1 | - | - | 1058 | 97 | |
AY064175.1 | - | 7 - 8 | 937 | 97 | |
BC011272.1 | - | 12 - 13 | 1758 | 97 | |
BC036957.1 | - | 11 - 12 | 1468 | 97 | |
M77842.1 | - | 7 - 8 | 1057 | 97 | |
X63963.1 | - | 12 - 13 | 1391 | 97 |