Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290549.1 | NP_001277478.1 | 8 - 9 | 1532 | 76 |
NM_001290551.1 | NP_001277480.1 | 7 - 8 | 1379 | 76 | |
NM_013690.3 | NP_038718.2 | 8 - 9 | 1532 | 76 | |
GenBank mRNA | BC050824.1 | - | 7 - 8 | 1373 | 76 |
D13738.1 | - | 8 - 9 | 1534 | 76 | |
S67051.1 | - | 9 - 10 | 1195 | 76 | |
X67553.1 | - | 8 - 9 | 1303 | 76 | |
X71426.1 | - | 9 - 10 | 1266 | 76 |