Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013708.4 | NP_038736.1 | 3 - 4 | 526 | 66 |
GenBank mRNA | AF148129.1 | - | 3 - 4 | 388 | 66 |
AF204053.1 | - | 3 - 4 | 526 | 66 | |
AK053560.1 | - | 3 - 4 | 494 | 66 | |
BC017521.1 | - | 3 - 4 | 520 | 66 |