Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289898.1 | NP_001276827.1 | 9 - 10 | 1357 | 62 |
NM_013767.6 | NP_038795.3 | 9 - 10 | 1361 | 62 | |
GenBank mRNA | AB028241.1 | - | 9 - 10 | 1307 | 62 |
AB028736.2 | - | 9 - 10 | 1307 | 62 | |
AK035978.1 | - | 3 - 4 | 457 | 62 | |
BC026127.1 | - | 9 - 10 | 1310 | 62 |