Assay Design



Gene Transcripts

Gene Symbol
Hk2
Entrez Gene ID
15277
Gene Name
hexokinase 2
Gene Aliases
AI642394, HKII
Chromosome Location
Chr.6: 82725025 - 82774454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013820.3 NP_038848.1 3 - 4 845 68
GenBank mRNA AJ238540.1 - 3 - 4 788 68
AK145936.1 - 3 - 4 826 68
BC054472.1 - 3 - 4 764 68

View Full Product Details