Assay Design



Gene Transcripts

Gene Symbol
Ndrg3
Entrez Gene ID
29812
Gene Name
N-myc downstream regulated gene 3
Gene Aliases
4833415O14Rik, AI747193, Ndr3
Chromosome Location
Chr.2: 156927342 - 156992111 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013865.2 NP_038893.1 13 - 14 1019 79
NM_180956.1 NP_851287.1 13 - 14 1019 79
GenBank mRNA AB033922.1 - 13 - 14 933 79
AK004689.1 - 13 - 14 965 79
AK037487.1 - 13 - 14 945 79
AK154927.1 - 13 - 14 986 79
AK185967.1 - 3 - 4 159 79
BC018504.1 - 13 - 14 953 79

View Full Product Details