Assay Design



Gene Transcripts

Gene Symbol
Fbxw4
Entrez Gene ID
30838
Gene Name
F-box and WD-40 domain protein 4
Gene Aliases
Dac, Fbw4, SHFM3, SHSF3, dactylin, dactylyn
Chromosome Location
Chr.19: 45578254 - 45660654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013907.2 NP_038935.1 7 - 8 1137 82
GenBank mRNA AB022163.1 - 7 - 8 1137 82
AF176519.1 - 7 - 8 1114 82
BC027031.1 - 7 - 8 999 82

View Full Product Details