Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293667.1 | NP_001280596.1 | 1 - 2 | 340 | 98 |
NM_001293668.1 | NP_001280597.1 | 1 - 2 | 340 | 98 | |
NM_015804.3 | NP_056619.1 | 1 - 2 | 340 | 98 | |
NR_121603.1 | 1 - 2 | 340 | 98 | ||
GenBank mRNA | AF156551.1 | - | 1 - 2 | 132 | 98 |
AK147497.1 | - | 1 - 2 | 343 | 98 | |
AK173088.1 | - | 1 - 2 | 287 | 98 | |
BC138715.1 | - | 1 - 2 | 59 | 98 | |
BC145399.1 | - | 1 - 2 | 60 | 98 | |
BC145400.1 | - | 1 - 2 | 60 | 98 |