Assay Design



Gene Transcripts

Gene Symbol
Atp11a
Entrez Gene ID
50770
Gene Name
ATPase, class VI, type 11A
Gene Aliases
4930558F19Rik, AU040868, Atpc1h, Ih
Chromosome Location
Chr.8: 12757016 - 12868728 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001293667.1 NP_001280596.1 13 - 14 1693 79
NM_001293668.1 NP_001280597.1 13 - 14 1693 79
NM_015804.3 NP_056619.1 13 - 14 1693 79
NR_121603.1 13 - 14 1693 79
GenBank mRNA AF156551.1 - 13 - 14 1485 79
AK147497.1 - 13 - 14 1696 79
AK173088.1 - 13 - 14 1846 79
BC138715.1 - 13 - 14 1412 79
BC145399.1 - 13 - 14 1413 79
BC145400.1 - 13 - 14 1413 79

View Full Product Details