Assay Design



Gene Transcripts

Gene Symbol
Syt5
Entrez Gene ID
53420
Gene Name
synaptotagmin V
Gene Aliases
SytV
Chromosome Location
Chr.7: 4539765 - 4547594 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016908.2 NP_058604.1 3 - 4 629 77
GenBank mRNA AB026806.1 - 3 - 4 373 77
BC047148.1 - 3 - 4 629 77

View Full Product Details