Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017391.3 | NP_059087.2 | 2 - 2 | 1649 | 68 |
GenBank mRNA | AK133628.1 | - | 1 - 1 | 2855 | 68 |
AK144718.1 | - | 2 - 2 | 1650 | 68 | |
BC140982.1 | - | 1 - 1 | 1196 | 68 | |
BC140983.1 | - | 1 - 1 | 1196 | 68 |