Assay Design



Gene Transcripts

Gene Symbol
Arhgef7
Entrez Gene ID
54126
Gene Name
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location
Chr.8: 11728105 - 11835219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 4 - 5 614 99
NM_001113518.1 NP_001106990.1 3 - 4 403 99
NM_017402.4 NP_059098.2 3 - 4 403 99
GenBank mRNA AF247654.1 - 3 - 4 299 99
AF343877.1 - 3 - 4 283 99
AK052545.1 - 3 - 4 374 99
AK129064.1 - 4 - 5 550 99
AK139156.1 - 4 - 5 543 99
AY220301.1 - 3 - 4 368 99
BC044838.1 - 3 - 4 388 99
U96634.1 - 1 - 2 93 99

View Full Product Details