Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019454.3 | NP_062327.2 | 10 - 11 | 2412 | 71 |
GenBank mRNA | AF253469.1 | - | 10 - 11 | 2367 | 71 |
AF273454.1 | - | 10 - 11 | 2430 | 70 | |
AK004739.1 | - | 10 - 11 | 2380 | 71 | |
AK031699.1 | - | 10 - 11 | 2382 | 71 | |
AK052322.1 | - | 10 - 11 | 2414 | 71 | |
AK165517.1 | - | 10 - 11 | 2380 | 71 | |
AK171717.1 | - | 10 - 11 | 2379 | 71 | |
BC042497.1 | - | 10 - 11 | 2367 | 71 | |
BC049130.1 | - | 10 - 11 | 2372 | 71 |