Assay Design



Gene Transcripts

Gene Symbol
Uso1
Entrez Gene ID
56041
Gene Name
USO1 vesicle docking factor
Gene Aliases
115kDa, TAP, Vdp
Chromosome Location
Chr.5: 92137909 - 92202800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019490.1 NP_062363.1 14 - 15 1721 106
GenBank mRNA AF096868.1 - 13 - 14 1509 106
AK144638.1 - 14 - 15 1712 106
AK156070.1 - 14 - 15 1714 106
BC016069.1 - 14 - 15 1721 106

View Full Product Details