Assay Design



Gene Transcripts

Gene Symbol
Uso1
Entrez Gene ID
56041
Gene Name
USO1 vesicle docking factor
Gene Aliases
115kDa, TAP, Vdp
Chromosome Location
Chr.5: 92137909 - 92202800 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019490.1 NP_062363.1 16 - 17 2016 151
GenBank mRNA AF096868.1 - 15 - 16 1804 151
AK156070.1 - 16 - 17 2009 151
AK168433.1 - 4 - 5 547 151
BC016069.1 - 16 - 17 2016 151

View Full Product Details