Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019490.1 | NP_062363.1 | 18 - 19 | 2288 | 62 |
GenBank mRNA | AF096868.1 | - | 17 - 18 | 2076 | 62 |
AK156070.1 | - | 18 - 19 | 2281 | 62 | |
AK168433.1 | - | 6 - 7 | 819 | 62 | |
BC005548.1 | - | 2 - 3 | 256 | 62 | |
BC016069.1 | - | 18 - 19 | 2288 | 62 |