Assay Design



Gene Transcripts

Gene Symbol
Fetub
Entrez Gene ID
59083
Gene Name
fetuin beta
Gene Aliases
2310011O17Rik, AI255764, D17980, Gugu
Chromosome Location
Chr.16: 22918382 - 22939768 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021564.2 NP_067539.1 1 - 2 273 97
GenBank mRNA AJ242927.1 - 1 - 2 194 97
AY138459.1 - 1 - 2 258 97

View Full Product Details