Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083904.1 | NP_001077373.1 | 5 - 6 | 737 | 68 |
NM_001083905.1 | NP_001077374.1 | 6 - 7 | 907 | 68 | |
NM_021564.2 | NP_067539.1 | 6 - 7 | 972 | 68 | |
GenBank mRNA | AJ242927.1 | - | 6 - 7 | 893 | 68 |
AK009310.1 | - | 6 - 7 | 962 | 68 | |
AK009554.1 | - | 6 - 7 | 906 | 68 | |
AK037043.1 | - | 6 - 7 | 908 | 68 | |
AY138459.1 | - | 6 - 7 | 957 | 68 | |
AY138460.1 | - | 5 - 6 | 507 | 68 | |
BC018341.1 | - | 6 - 7 | 837 | 68 |