Assay Design



Gene Transcripts

Gene Symbol
Retn
Entrez Gene ID
57264
Gene Name
resistin
Gene Aliases
ADSF, Fizz3, Rstn, Xcp4
Chromosome Location
Chr.8: 3655770 - 3659818 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204959.1 NP_001191888.1 1 - 2 74 93
NM_022984.4 NP_075360.1 1 - 2 74 93
GenBank mRNA AF290870.1 - 1 - 2 67 93
AF323080.1 - 1 - 2 73 93
AK131613.1 - 1 - 2 75 93
AK131869.1 - 1 - 2 71 93
BC051196.1 - 1 - 2 60 93

View Full Product Details