Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023179.3 | NP_075668.1 | 2 - 3 | 349 | 103 |
GenBank mRNA | AB076405.1 | - | 2 - 3 | 404 | 103 |
AK005109.1 | - | 2 - 3 | 229 | 102 | |
AK011945.1 | - | 2 - 3 | 192 | 103 | |
BC020190.1 | - | 2 - 3 | 345 | 103 | |
BC049587.1 | - | 2 - 3 | 214 | 103 | |
BC062380.1 | - | 2 - 3 | 337 | 103 | |
BU558667.1 | - | 2 - 3 | 352 | 103 |