Assay Design



Gene Transcripts

Gene Symbol
Ndufa13
Entrez Gene ID
67184
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13
Gene Aliases
2700054G14Rik, AU022060, CDA016, CGI-39, GRIM-19, Grim19
Chromosome Location
Chr.8: 69887788 - 69902558 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023312.2 NP_075801.1 2 - 3 870 68
GenBank mRNA AF286698.1 - 1 - 2 105 68
AK002999.1 - 1 - 2 109 68
AK003012.1 - 1 - 2 123 68
AK010517.1 - 1 - 2 109 68
AK012422.1 - 1 - 2 109 68
BC034899.1 - 2 - 3 870 68
BC037149.1 - 1 - 2 113 68
BC055435.1 - 1 - 2 119 68

View Full Product Details