Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027324.5 | NP_081600.1 | 2 - 3 | 262 | 60 |
GenBank mRNA | AF325260.1 | - | 2 - 3 | 240 | 60 |
AK012650.1 | - | 2 - 3 | 261 | 60 | |
AK151110.1 | - | 2 - 3 | 208 | 60 | |
AK179717.1 | - | 1 - 2 | 47 | 60 | |
BC005743.1 | - | 2 - 3 | 219 | 60 |