Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033622.1 | NP_296371.1 | 4 - 5 | 760 | 79 |
GenBank mRNA | AF119383.1 | - | 4 - 5 | 760 | 79 |
AF352245.1 | - | 4 - 5 | 764 | 79 | |
AK034121.1 | - | 4 - 5 | 392 | 79 | |
AK155288.1 | - | 4 - 5 | 539 | 79 | |
BC106841.1 | - | 4 - 5 | 733 | 79 |