Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033622.1 | NP_296371.1 | 6 - 7 | 1035 | 66 |
GenBank mRNA | AF119383.1 | - | 6 - 7 | 1035 | 66 |
AK034121.1 | - | 6 - 7 | 667 | 66 | |
AK079180.1 | - | 6 - 7 | 1254 | 66 | |
AK155288.1 | - | 6 - 7 | 814 | 66 | |
AY290823.1 | - | 5 - 6 | 762 | 66 | |
BC106840.1 | - | 5 - 6 | 951 | 66 | |
BC106841.1 | - | 6 - 7 | 1008 | 66 |