Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290389.1 | NP_001277318.1 | 6 - 7 | 944 | 82 |
NM_133690.3 | NP_598451.1 | 6 - 7 | 956 | 82 | |
GenBank mRNA | AF348324.1 | - | 6 - 7 | 811 | 82 |
AF348325.1 | - | 6 - 7 | 799 | 82 | |
AK053500.1 | - | 6 - 7 | 895 | 82 | |
BC137923.1 | - | 6 - 7 | 897 | 82 | |
BC137924.1 | - | 6 - 7 | 909 | 82 |