Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133977.2 | NP_598738.1 | 8 - 9 | 1118 | 62 |
GenBank mRNA | AF440692.1 | - | 7 - 8 | 1048 | 62 |
AK085754.1 | - | 8 - 9 | 1103 | 62 | |
AK142599.1 | - | - | 1108 | 62 | |
AK146549.1 | - | 8 - 9 | 1103 | 62 | |
AK149559.1 | - | 8 - 9 | 1103 | 62 | |
AK149595.1 | - | 8 - 9 | 1103 | 62 | |
AK150782.1 | - | 8 - 9 | 1151 | 62 | |
AK168368.1 | - | 8 - 9 | 1105 | 62 | |
AK168405.1 | - | 8 - 9 | 1105 | 62 | |
AK168419.1 | - | 8 - 9 | 1103 | 62 | |
AK168938.1 | - | 8 - 9 | 1103 | 62 | |
AK169187.1 | - | 8 - 9 | 1102 | 62 | |
BC008559.1 | - | 6 - 7 | 840 | 62 | |
BC012313.1 | - | 8 - 9 | 1118 | 62 | |
BC018573.1 | - | 3 - 4 | 388 | 62 | |
BC020295.1 | - | 8 - 9 | 1087 | 62 | |
BC022986.1 | - | 8 - 9 | 1096 | 62 | |
BC058216.1 | - | 8 - 9 | 1084 | 62 | |
BC058218.1 | - | 8 - 9 | 1097 | 62 | |
BC092046.1 | - | 8 - 9 | 1079 | 62 |