Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293313.1 | NP_001280242.1 | 7 - 8 | 795 | 78 |
NM_138314.4 | NP_612187.2 | 7 - 8 | 798 | 78 | |
NM_178071.6 | NP_835172.1 | 7 - 8 | 795 | 78 | |
GenBank mRNA | AF202048.1 | - | 6 - 7 | 699 | 78 |
AK044962.1 | - | 7 - 8 | 769 | 78 | |
AK085222.1 | - | 7 - 8 | 743 | 78 | |
AK133221.1 | - | - | 1266 | 78 | |
AK144915.1 | - | 7 - 8 | 799 | 78 | |
AK164652.1 | - | 7 - 8 | 743 | 78 | |
BC038021.1 | - | 8 - 9 | 78 | ||
BC049225.1 | - | 6 - 7 | 78 |