Assay Design
Gene Transcripts
- Gene Symbol
- Sec14l2
- Entrez Gene ID
-
67815
- Gene Name
- SEC14-like 2 (S. cerevisiae)
- Gene Aliases
-
1300013M05Rik, Spf, TAP
- Chromosome Location
-
Chr.11: 4097039 - 4118729 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_144520.2 |
NP_653103.1 |
6 - 7 |
573 |
71 |
GenBank mRNA |
BC005759.1 |
- |
6 - 7 |
573 |
71 |
View Full Product Details