Assay Design



Gene Transcripts

Gene Symbol
Pou3f4
Entrez Gene ID
18994
Gene Name
POU domain, class 3, transcription factor 4
Gene Aliases
Brn-4, Brn4, OTF-9, Otf9, Slf, oct-9
Chromosome Location
Chr.X: 110814465 - 110815550 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008901.1 NP_032927.1 1 - 1 350 65
GenBank mRNA AK158302.1 - 1 - 1 383 65
BC052012.1 - 1 - 1 398 65
BC086670.1 - - 361 65
BC138657.1 - 1 - 1 425 65
BC145919.1 - 1 - 1 425 65
X66603.1 - - 393 65

View Full Product Details