Assay Design



Gene Transcripts

Gene Symbol
Ppl
Entrez Gene ID
19041
Gene Name
periplakin
Gene Aliases
AW553870
Chromosome Location
Chr.16: 5086291 - 5132481 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008909.2 NP_032935.2 7 - 8 899 68
GenBank mRNA AF126834.1 - 7 - 8 839 68
AK142562.1 - 7 - 8 894 68
BC057152.1 - 7 - 8 828 68

View Full Product Details