Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042451.2 | NP_001035916.1 | 4 - 5 | 618 | 74 |
NM_009221.2 | NP_033247.1 | 4 - 5 | 522 | 74 | |
GenBank mRNA | AF033261.1 | - | 4 - 5 | 445 | 74 |
AF044672.1 | - | 3 - 4 | 350 | 74 | |
AF179273.1 | - | 4 - 5 | 522 | 74 | |
AK156316.1 | - | 4 - 5 | 454 | 74 | |
BC046764.1 | - | 4 - 5 | 361 | 74 |