Assay Design



Gene Transcripts

Gene Symbol
Trim21
Entrez Gene ID
20821
Gene Name
tripartite motif-containing 21
Gene Aliases
Ro52, Ssa1
Chromosome Location
Chr.7: 102557920 - 102565482 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009277.4 NP_033303.3 1 - 2 92 71
GenBank mRNA AK141294.1 - 1 - 2 71 71
AK171489.1 - 1 - 2 56 71
BB661457.1 - 1 - 2 79 71
CJ051157.1 - 1 - 2 94 71
L27990.1 - 1 - 2 57 71

View Full Product Details