Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009396.2 | NP_033422.2 | 3 - 4 | 1006 | 82 |
GenBank mRNA | AK087680.1 | - | 3 - 4 | 1005 | 82 |
AK154405.1 | - | 3 - 4 | 1006 | 82 | |
AK170719.1 | - | 3 - 4 | 1006 | 82 | |
AK172653.1 | - | 3 - 4 | 1006 | 82 | |
BC138106.1 | - | 2 - 3 | 915 | 82 | |
BC138107.1 | - | 2 - 3 | 915 | 82 | |
L24118.1 | - | 2 - 3 | 950 | 82 |