Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159401.1 | NP_001152873.1 | 4 - 5 | 420 | 70 |
NM_001159402.1 | NP_001152874.1 | 4 - 5 | 419 | 70 | |
NM_009477.3 | NP_033503.2 | 4 - 5 | 447 | 70 | |
GenBank mRNA | AK136474.1 | - | 4 - 5 | 300 | 70 |
AL118479.1 | - | 4 - 5 | 420 | 70 | |
AV447018.1 | - | 4 - 5 | 471 | 70 | |
BC036559.1 | - | 4 - 5 | 321 | 70 | |
CA493594.1 | - | - | 457 | 70 | |
CA536983.1 | - | 4 - 5 | 424 | 70 | |
D44464.1 | - | 4 - 5 | 448 | 70 |