Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011078.3 | NP_035208.2 | 15 - 16 | 2218 | 110 |
GenBank mRNA | AB221590.1 | - | 4 - 5 | 604 | 110 |
AF043726.1 | - | 15 - 16 | 2218 | 110 | |
AK172995.1 | - | 13 - 14 | 1925 | 110 | |
BC051633.1 | - | 15 - 16 | 2216 | 110 |