Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011201.3 | NP_035331.3 | 6 - 7 | 864 | 142 |
GenBank mRNA | AK149979.1 | - | 6 - 7 | 820 | 142 |
AK150322.1 | - | 6 - 7 | 820 | 142 | |
AK151371.1 | - | 6 - 7 | 777 | 142 | |
AK153088.1 | - | 6 - 7 | 802 | 142 | |
AK157431.1 | - | 6 - 7 | 865 | 142 | |
AK165949.1 | - | 7 - 8 | 944 | 142 | |
AK166020.1 | - | 6 - 7 | 820 | 142 | |
AK168255.1 | - | 6 - 7 | 802 | 142 | |
AK168772.1 | - | 6 - 7 | 819 | 142 | |
AK170620.1 | - | 6 - 7 | 820 | 142 | |
AK171380.1 | - | 6 - 7 | 820 | 142 | |
AK172206.1 | - | 7 - 8 | 937 | 142 | |
BC005729.1 | - | 1 - 2 | 217 | 142 | |
BC010191.1 | - | 6 - 7 | 782 | 142 | |
M97590.1 | - | 6 - 7 | 780 | 142 |