Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166381.1 | NP_001159853.1 | 7 - 8 | 693 | 149 |
NM_001166382.1 | NP_001159854.1 | 7 - 8 | 693 | 149 | |
NM_001166383.1 | NP_001159855.1 | 7 - 8 | 693 | 149 | |
NM_011236.2 | NP_035366.2 | 7 - 8 | 693 | 149 | |
GenBank mRNA | AK080702.1 | - | 7 - 8 | 666 | 149 |
BC010673.1 | - | 6 - 7 | 565 | 149 | |
BC019123.1 | - | 7 - 8 | 673 | 149 | |
U06837.1 | - | 7 - 8 | 615 | 149 | |
U12135.1 | - | 7 - 8 | 662 | 149 | |
Z32767.1 | - | 7 - 8 | 687 | 149 |