Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011960.2 | NP_036090.2 | 1 - 2 | 490 | 82 |
GenBank mRNA | AF079557.1 | - | 1 - 2 | 239 | 82 |
AK036656.1 | - | 1 - 2 | 513 | 82 | |
AK163388.1 | - | 1 - 2 | 514 | 82 | |
AK164013.1 | - | 1 - 2 | 477 | 82 | |
BC050892.1 | - | 1 - 2 | 489 | 82 | |
BC053037.1 | - | 1 - 2 | 473 | 82 | |
BC059827.1 | - | 1 - 2 | 499 | 82 |