Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012038.4 | NP_036168.1 | 2 - 3 | 457 | 72 |
GenBank mRNA | AJ489331.1 | - | 2 - 3 | 421 | 72 |
AK144454.1 | - | 2 - 3 | 387 | 72 | |
AK160224.1 | - | 2 - 3 | 389 | 72 | |
AY101375.1 | - | 1 - 2 | 159 | 72 | |
BC046226.1 | - | 2 - 3 | 362 | 72 | |
D21165.1 | - | 1 - 2 | 159 | 72 | |
EU373446.1 | - | 1 - 2 | 159 | 72 |