Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001009978.1 | NP_001009978.1 | 4 - 5 | 418 | 97 |
NM_001009979.1 | NP_001009979.1 | 4 - 5 | 418 | 97 | |
NM_001159582.1 | NP_001153054.1 | 6 - 7 | 920 | 97 | |
NM_016744.4 | NP_058024.2 | 5 - 6 | 715 | 97 | |
GenBank mRNA | AF023529.1 | - | 5 - 6 | 635 | 97 |
AF159298.1 | - | 4 - 5 | 418 | 97 | |
AK014810.1 | - | 4 - 5 | 417 | 97 | |
AK043647.1 | - | 5 - 6 | 716 | 97 | |
AK050004.1 | - | 6 - 7 | 781 | 97 | |
AY845863.1 | - | 5 - 6 | 467 | 97 | |
AY845864.1 | - | 4 - 5 | 418 | 97 | |
BC090628.1 | - | 5 - 6 | 683 | 97 | |
U56649.1 | - | 6 - 7 | 651 | 97 |